Learn More
STEMCELL Technologies Mouse Recombinant SCF
Artikelnummer. 17159481
Encompass_Preferred
Click to view available options
Menge:
10 μg
100 μg
1000 μg
500 μg
Packungsgröße:
10 Mikrogramm
500 Mikrogramm
Artikelnummer. 17159481
Marke: STEMCELL Technologies 78064
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Stem cell factor
  • Stem cell factor (SCF) is an early-acting cytokine that plays a pivotal role in the regulation of embryonic and adult hematopoiesis
  • SCF promotes cell survival, proliferation, differentiation, adhesion, and functional activation of cells at multiple levels of the hematopoietic hierarchy
  • Together with other cytokines such as thrombopoietin and Flt3/Flk-2 Ligand, SCF is commonly used to promote expansion of primitive hematopoietic stem cells and multipotent progenitor cells in culture
  • In synergy with various growth factors, including IL-2, IL-3, IL-6, IL-7, G-CSF, and erythropoietin, SCF increases proliferation and differentiation of myeloid and erythroid progenitor cells and a subset of lymphoid progenitor cells
  • It is produced by stromal cells in the fetal liver, bone marrow, and thymus, in the central nervous system, in keratinocytes, and in the gut mucosa, and can function as a chemotactic and chemokinetic factor
Spezifikation
Name Kit ligand, Mast cell growth factor, Steel factor, Stem cell factor
Menge 100 μg
Zelltyp Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells
Reinheits- oder Qualitätsgrad ≥95%