Learn More
STEMCELL Technologies Mouse Recombinant VEGF-164
Click to view available options
Menge:
100 μg
1000 μg
20 μg
Packungsgröße:
1000 Mikrogramm
Beschreibung
- Vascular endothelial growth factor (VEGF) is a heparin-binding homodimeric glycoprotein involved in vasculogenesis and angiogenesis
- VEGF binds to FLT1 (VEGFR-1) and KDR (VEGFR-2), and activates Raf/MEK/ERK and PI3K/AKT pathways
- VEGF plays an important role in neurogenesis both in vitro and in vivo
- It has neurotrophic effects on neurons of the central nervous system, and it promotes growth and survival of dopaminergic neurons and astrocytes
- VEGF also promotes growth and survival of vascular endothelial cells, monocyte chemotaxis, and colony formation by granulocyte-macrophage progenitor cells
Spezifikation
Spezifikation
| Name | MGC70609, MVCD-1, Vascular endothelial growth factor 2, Vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VPF |
| Menge | 1000 μg |
| Zelltyp | Angiogenic Cells; Endothelial Cells; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells |
| Reinheits- oder Qualitätsgrad | ≥97% |