missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse Siglec-15 / CD33L3 Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_001094508.1 |
---|---|
Molekulargewicht | 27.5 kDa |
Quelle | Mouse Siglec-15, His Tag (SG5-M52H7) is expressed from human 293 cells (HEK293). It contains AA Arg 24 - Thr 262 (Accession # A7E1W8-1). |
Immunogen | Arg 24 - Thr 262 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16448039
|
ACROBiosystems
SG5-M52H7-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16438039
|
ACROBiosystems
SG5-M52H7-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Siglec-15 is a DAP12-associated immunoreceptor, which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. Siglecs are cell surface proteins that bind sialic acid. They are found primarily on the surface of immune cells and are a subset of the I-type lectins. Siglec-15 consisting of immunoglobulin (Ig)-like domains, transmembrane domain and a short cytoplasmic tail. Siglec-15 is that recognizes sialylated glycans and regulates osteoclast differentiation. Siglec-15 is a potential therapeutic target for osteoporosis and plays a conserved regulatory role in the immune system of vertebrates.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.5 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001094508.1 | |
Mouse Siglec-15, His Tag (SG5-M52H7) is expressed from human 293 cells (HEK293). It contains AA Arg 24 - Thr 262 (Accession # A7E1W8-1). | |
-20°C | |
Siglec-15 | |
Mouse | |
PBS with Arginine, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
27.5 kDa | |
Arg 24 - Thr 262 | |
CD33 antigen-like 3,SIGLEC-15,CD33L3,sialic acid-binding Ig-like lectin 15,Siglec15,Siglec-15 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |