Learn More
ACROBiosystems Mouse Siglec-2 / CD22 Protein, His Tag
362.89 € - 2661.24 €
Spezifikation
Zugriffsnummer | P35329.1 |
---|---|
Molekulargewicht | 78.3 kDa |
Quelle | Mouse Siglec-2, His Tag (SI2-M52Ha) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 702 (Accession # P35329-1). |
Immunogen | Ser 22 - Arg 702 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16428059
|
ACROBiosystems
SI2-M52HA-100UG |
100 μg |
362.89 €
100 Mikrogramm |
Verfügbar ab: 27-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16438059
|
ACROBiosystems
SI2-M52HA-1MG |
1 mg |
2661.24 €
1 Milligramm |
Verfügbar ab: 27-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
B-cell receptor CD22 is also known as Sialic acid-binding Ig-like lectin 2 (Siglec-2), B-lymphocyte cell adhesion molecule (BL-CAM), T-cell surface antigen Leu-14, which belongs to the immunoglobulin superfamily and SIGLEC (sialic acid binding Ig-like lectin) family. CD22 mediates B-cell B-cell interactions, and may be involved in the localization of B-cells in lymphoid tissues. Siglec-2 / CD22 binds sialylated glycoproteins, one of which is CD45. Siglec2 / CD22 plays a role in positive regulation through interaction with Src family tyrosine kinases and may also act as an inhibitory receptor by recruiting cytoplasmic phosphatases via their SH2 domains that block signal transduction through dephosphorylation of signaling molecules.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 78.3 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
P35329.1 | |
Mouse Siglec-2, His Tag (SI2-M52Ha) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 702 (Accession # P35329-1). | |
-20°C | |
Siglec-2 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
78.3 kDa | |
Ser 22 - Arg 702 | |
CD22,SIGLEC2,BL-CAM,SIGLEC-2,Siglec2,SIGLEC2FLJ22814 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |