Learn More
ACROBiosystems Mouse TIGIT Protein, Fc Tag
423.38 € - 2661.24 €
Spezifikation
Zugriffsnummer | NP_001139797.1 |
---|---|
Molekulargewicht | 39.5 kDa |
Quelle | Mouse TIGIT, Fc Tag (TIT-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # NP_001139797.1). |
Immunogen | Gly 26 - Thr 143 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15847787
|
ACROBiosystems
TIT-M5257-1MG |
1 mg |
2661.24 €
1 Milligramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15857787
|
ACROBiosystems
TIT-M5257-50UG |
50 μg |
423.38 €
50 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
T-cell immunoreceptor with Ig and ITIM domains (TIGIT) is also known as V-set and immunoglobulin domain-containing protein 9 (VSIG9), V-set and transmembrane domain-containing protein 3 (VSTM3),which belongs to single-pass type I membrane protein containing an immunoglobulin variable domain, a transmembrane domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM). TIGIT is expressed at low levels on peripheral memory and regulatory CD4+ T-cells and NK cells and is up-regulated following activation of these cells (at protein level). TIGIT binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T-cell activation by promoting the generation of mature immunoregulatory dendritic cells.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.5 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_001139797.1 | |
Mouse TIGIT, Fc Tag (TIT-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession # NP_001139797.1). | |
-20°C | |
TIGIT | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
39.5 kDa | |
Gly 26 - Thr 143 | |
TIGIT,VSIG9,VSTM3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |