Learn More
MOXD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557381
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DINDKSITDS TVRVIWAYHH EDAGEAGPKY HDSNRGTKSL RLLNPEKTSV LSTALPYFDL VNQDVPIPNK DTTYWCQMFK IPVFQEKHHV IKVEPV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 90%.
MOXD1 Polyclonal antibody specifically detects MOXD1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
MOXD1 | |
Polyclonal | |
Unconjugated | |
MOXD1 | |
3230402N08Rik; DBH-like monooxygenase protein 1; Dbhr; DBH-related protein; dJ248E1.1; dopamine-oxygenase; MNCb-5203; monooxygenase DBH like 1; monooxygenase X; monooxygenase, DBH-like 1; MOX; MOXD1; PRO5780; UNQ2493/PRO5780 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26002 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UVY6 | |
MOXD1 | |
Recombinant protein corresponding to Human MOXD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only