Learn More
MPP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529311
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, mouse brain. Predicted reactivity: Mouse (97%), Rat (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MPP2 Polyclonal antibody specifically detects MPP2 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
MPP2 | |
Polyclonal | |
Unconjugated | |
Mpp2 | |
D11Bwg0652e; discs large homolog 2; discs large, homolog 2; Dlg2; Dlgh2; MAGUK p55 subfamily member 2; membrane palmitoylated protein 2; membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2); Mpp2; p55 subfamily member of MAGUK family; palmitoylated membrane protein 2; Pals4; Protein MPP2; rCG_34691 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4355, 50997 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q14168, Q9WV34 | |
Mpp2 | |
Recombinant fragment corresponding to a region within amino acids 297 and 545 of Human MPP2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |