Learn More
MRPL27 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554402
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HPGAHVGVGK NKCLYALEEG IVRYTKEVYV PHPRNTEAVD LITRLPKGAV LYKTFVHVVP AKPEGTFKLV AML Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
MRPL27 Polyclonal antibody specifically detects MRPL27 in Human samples. It is validated for Immunohistochemistry.Spezifikation
MRPL27 | |
Polyclonal | |
Unconjugated | |
MRPL27 | |
39S ribosomal protein L27, mitochondrial; A630055F16Rik; D11Moh47; D18Ertd643e; HSPC250; L27mt; Mitochondrial large ribosomal subunit protein bL27m; mitochondrial ribosomal protein L27; Mrpl27; MRP-L27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51264 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0M9 | |
MRPL27 | |
Recombinant protein corresponding to Human MRPL27 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only