Learn More
MRPL57 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558926
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RFVSLRAKQN MIRRLEIEAE NHYWLSMPYM TREQERGHAA VRRREAFEAI KA Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 81%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a protein which belongs to an undetermined ribosomal subunit and which seems to be specific to animal mitoribosomes. Pseudogenes corresponding to this gene are found on chromosomes 1p, 1q, 3p, 5q, 8q, 14q, and Y.Spezifikation
MRPL57 | |
Polyclonal | |
Unconjugated | |
MRPL57 | |
bMRP63; hMRP63; Mitochondrial large ribosomal subunit protein mL63; Mitochondrial ribosomal protein 63; mitochondrial ribosomal protein bMRP63; mitochondrial ribosomal protein L57; MRP63; MRPL57; Ribosomal protein 63, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
78988 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BQC6 | |
MRPL57 | |
Recombinant protein corresponding to Human MRPL57 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only