missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS26 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15989-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. This gene lies adjacent to and downstream of the gonadotropin-releasing hormone precursor gene.Spezifikation
MRPS26 | |
Polyclonal | |
Unconjugated | |
MRPS26 | |
C20orf193, GI008, MRP S13, MRP S26, MRPS13, MRPS26, NY BR 87, RPMS13, S13mt, S26mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362216, 64949, 99045 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80ZS3, Q9BYN8, Q9EPJ3 | |
Mrps26 | |
MRPS26 Fusion Protein Ag8797 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |