Learn More
MS4A6A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552826
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SFSPNFTQVT STLLNSAYPF IGPFFVSRVS EEGRMGQRGE EDTNSLDFPP ASLLCLICQE QGVNGESCSP V Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 25%.
May be involved in signal transduction as a component of a multimeric receptor complex.Spezifikation
MS4A6A | |
Polyclonal | |
Unconjugated | |
MS4A6A | |
4SPAN3; 4SPAN3.2; CD20 antigen-like 3; CD20L3; CD20-like precusor; CDA01; four-span transmembrane protein 3; four-span transmembrane protein 3.1; four-span transmembrane protein 3.2; HAIRB-iso; membrane spanning 4-domains A6A; membrane-spanning 4-domains subfamily A member 6A; membrane-spanning 4-domains subfamily A member 6D; membrane-spanning 4-domains, subfamily A, member 11; membrane-spanning 4-domains, subfamily A, member 6A; membrane-spanning 4-domains, subfamily A, member 6A, isoform 2; Ms4a11; MS4A6; MS4A6A; MS4A6A-polymorph; MST090; MSTP090 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2W1 | |
MS4A6A | |
Recombinant protein corresponding to Human MS4A6A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only