missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSI1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 27185-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Musashi-1 is a member of an evolutionarily conserved family of RNA-binding proteins that is prefentially expressed in the nervous system. It is a neural RNA-binding protein that is strongly expressed in fetal and adult neural stem cells (NSCs). Functionally, Musashi-1 plays important roles in cell fate decision, including the maintece of the stem-cell state, differentiation and tumorigenesis. Musashi-1 is an important regulator of the Notch-1 pathway.Spezifikation
MSI1 | |
Polyclonal | |
Unconjugated | |
MSI1 | |
MSI1, Musashi 1, musashi homolog 1 (Drosophila) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4440 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43347 | |
MSI1 | |
MSI1 Fusion Protein Ag26042 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |