Learn More
MT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552042
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MDPETCPCPS GGSCTCADSC KCEGCKCTSC KKSCCSCCPA ECEKCAKDCV CKGGEAAEAE AEKCS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.
MT3 Polyclonal antibody specifically detects MT3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
MT3 | |
Polyclonal | |
Unconjugated | |
MT3 | |
GIF; GIFB; GRIF; growth inhibitory factor; metallothionein 3; metallothionein 3 (growth inhibitory factor (neurotrophic)); metallothionein III; metallothionein-3; Metallothionein-III; MT3; MT-3; MT-III; ZnMT3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4504 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P25713 | |
MT3 | |
Recombinant protein corresponding to Human MT3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only