Learn More
MTHFD1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556046
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ARDSIVREVI QNSKEVLSLL QEKNPAFKPV LAIIQAGDDN LMQEINQNLA EEAGLNITHI CLPPDSSEAE IIDEILKINE DTRVHGLALQ Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
One-carbon substituted forms of tetrahydrofolate (THF) are involved in the de novo synthesis of purines and thymidylate and support cellular methylation reactions through the regeneration of methionine from homocysteine. MTHFD1L is an enzyme involved in THF synthesis in mitochondria (Christensen et al., 2005 [PubMed 15611115]).Spezifikation
MTHFD1L | |
Polyclonal | |
Unconjugated | |
MTHFD1L | |
10-formyl-THF synthetase; 2410004L15Rik; AI647056; dJ292B18.2; Formyltetrahydrofolate synthetase; formyltetrahydrofolate synthetase domain containing 1; FTHFSDC1; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like; monofunctional C1-tetrahydrofolate synthase, mitochondrial; MTC1THFS; MTHFD1L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25902 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UB35 | |
MTHFD1L | |
Recombinant protein corresponding to Human MTHFD1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only