Learn More
MTMR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556961
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MEPSPSETSL VERPQVGSVV HRTSLGSTLS LTRSPCALPL AECKEGLVCN GAPETENRAS EQPPGLSTLQ MYPTPNGHCA NGEAGRSKDS LSR Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 66%.
This gene encodes a member of the myotubularin dual specificity protein phosphatase gene family. The encoded protein is structurally similar to myotubularin but in addition contains a FYVE domain and an N-terminal PH-GRAM domain. The protein can self-associate and also form heteromers with another myotubularin related protein. The protein binds to phosphoinositide lipids through the PH-GRAM domain, and can hydrolyze phosphatidylinositol(3)-phosphate and phosphatidylinositol(3,5)-bisphosphate in vitro. The encoded protein has been observed to have a perinuclear, possibly membrane-bound, distribution in cells, but it has also been found free in the cytoplasm. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
MTMR3 | |
Polyclonal | |
Unconjugated | |
MTMR3 | |
1700092A20Rik; AI255150; AW557713; FYVE (Fab1 YGLO23 Vsp27 EEA1 domain) dual-specificity protein phosphatase; FYVE domain-containing dual specificity protein phosphatase 1; FYVE-DSP1; KIAA0371; mKIAA0371; MTMR3; myotubularin related protein 3; myotubularin-related protein 3; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphate phosphatase; ZFYVE10; zinc finger FYVE domain-containing protein 10; zinc finger, FYVE domain containing 10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13615 | |
MTMR3 | |
Recombinant protein corresponding to Human MTMR3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only