missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTMR7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550342
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total MTMR7 protein.
MTMR7 Polyclonal antibody specifically detects MTMR7 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
MTMR7 | |
Polyclonal | |
Unconjugated | |
Mtmr7 | |
DKFZp781E194; Inositol 1,3-bisphosphate phosphatase; MGC163449; MGC163451; MTMR7; myotubularin related protein 7; myotubularin-related protein 7; Phosphatidylinositol-3-phosphate phosphatase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9108 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y216 | |
Mtmr7 | |
Fusion protein corresponding to residues near the C terminal of human myotubularin related protein 7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |