Learn More
Mecmesin™ MULTITEST 5-I STAND 5000N 600MM N
Artikelnummer. 11933828
Encompass
Ansicht ändern
Click to view available options
Bereich:5000N
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
11933828 Bereich:5000N
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11933828 Lieferant Mecmesin™ Lieferanten-Nr. 805014