missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MVP Recombinant Rabbit Monoclonal Antibody (24GB465)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557453
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MVP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.
Spezifikation
| MVP | |
| Recombinant Monoclonal | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14764, Q62667, Q9EQK5 | |
| MVP | |
| A synthesized peptide derived from human MVP (aa 1-55). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB465 | |
| Unconjugated | |
| MVP | |
| 2310009M24Rik; LRP; Lung resistance-related protein; major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64681, 78388, 9961 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur