missing translation for 'onlineSavingsMsg'
Learn More

N-(3 -Bromophenyl)-4-Chlorobutyramid, 97 %, Thermo Scientific™

Catalog Number p-7039905
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
549544-24-3
C10H11BrClNO
276.558
MFCD01346441
GHKCNIGXORDORM-UHFFFAOYSA-N
n-3-bromophenyl-4-chlorobutanamide, n-3-bromophenyl-4-chlorobutyramide
4184874
N-(3-Bromphenyl)-4-chlorbutanamid
C1=CC(=CC(=C1)Br)NC(=O)CCCCl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
549544-24-3
C10H11BrClNO
276.558
MFCD01346441
GHKCNIGXORDORM-UHFFFAOYSA-N
n-3-bromophenyl-4-chlorobutanamide, n-3-bromophenyl-4-chlorobutyramide
4184874
N-(3-Bromphenyl)-4-chlorbutanamid
C1=CC(=CC(=C1)Br)NC(=O)CCCCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-(3-Bromophenyl)-4-chlorobutyramide
Menge 250 mg
Formelmasse 276.56
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'