missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Hydrazinyl-6-oxohexyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, 97%, BLD Pharm™

Product Code. 31024639
Encompass_Preferred
Change view
Click to view available options
Menge:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31024639 250 mg
31022479 100 mg
31019249 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31024639 Supplier BLD Pharm Supplier No. BD116732250mg
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material N-(6-Hydrazinyl-6-oxohexyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide
CAS 109276-34-8
Summenformel C16H29N5O3S
MDL-Nummer MFCD00077659
Menge 250 mg
SMILES O=C(CCCC[C@@H]1SC[C@]([H])(N2)[C@@]1(NC2=O)[H])NCCCCCC(NN)=O
Molekulargewicht (g/mol) 371.50
Reinheit (%) 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.