Learn More
Thermo Scientific™ N-Boc-Arg-Val-Arg-Arg-7-Amino-4-Methylcoumarin
für zell- und molekularbiologische Anwendungen.
Marke: Thermo Scientific™ J66336.#0
Weitere Details : CAS : 136132-77-9 Netto-Gewicht : 0.00005kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
HPLC | |
Fest | |
N-Boc-Arg-Val-Arg-Arg-7-amino-4-methylcoumarin | |
RUO - research use only |
-30°C to -10°C | |
843.00 | |
5 mg | |
Boc-R-V-R-R-AMC |
Nur für Forschungszwecke