missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-Glycin N-succinimidylester, 98 %, Thermo Scientific Chemicals

Catalog Number p-7041542
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3392-07-2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
3392-07-2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-glycine N-succinimidyl ester
Schmelzpunkt 165°C to 167°C
Summenformel C15H24N2O6
MDL-Nummer MFCD00022585 MFCD00022585
Menge 5 g
Beilstein 1549242
Empfindlichkeit Hygroscopic
Synonym boc-ile-osu, boc-l-isoleucine hydroxysuccinimide ester, boc-l-isoleucine n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoate, boc-lle-osu, boc-ile-osu hplc, n-alpha-t-butyloxycarbonyl-l-isoleucine succinimidyl ester, 1-n-1,1-dimethylethoxy carbonyl-l-isoleucyl oxy-2,5-pyrrolidinedione, 2,5-dioxopyrrolidin-1-yl 2s,3s-3-methyl-2-2-methylpropan-2-yl oxycarbonylamino pentanoate
InChI-Schlüssel FATJLEZSGFVHQA-CABZTGNLSA-N
SMILES CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
IUPAC-Name (2,5-Dioxopyrrolidin-1-yl) (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoat
Molekulargewicht (g/mol) 328.37
PubChem CID 131632401
Formelmasse 272.25
Reinheit (%) 98%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'