Learn More
N-cadherin Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech CL488-22018-100UL
Weitere Details : Netto-Gewicht : 0.00010kg
Artikelnummer. 17204686
  • 430.00 € / 100 Mikroliter
Verfügbar ab: 24-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Cadherins are a family of transmembrane glycoproteins that mediate calcium-dependent cell-cell adhesion and play an important role in the maintenance of normal tissue architecture. N-cadherin (neural cadherin), also known as CDH2 (cadherin 2), is a 130-kDa transmembrane glycoprotein and a classical member of the cadherin superfamily which also include E-, P-, R-, and B-cadherins. Expression of N-cadherin has been reported on various cell types including neurons, endothelial cells and cardiac myocytes (PMID: 11282032; 9508779; 8125202). N-cadherin has functions in early brain morphogenesis, synaptogenesis and synaptic plasticity (PMID: 23321619).
Spezifikation
Spezifikation
N-cadherin
Polyclonal
CoraLite 488
100 μL
1000
Liquid
Immunofluorescence
1000 μg/mL
Rabbit
Primary
Human, Mouse, Rat