missing translation for 'onlineSavingsMsg'
Learn More

N-Methylmethacrylamide, 98%, BLD Pharm™

Artikelnummer. 30987621
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30987621 1 g
30981624 25 g
30975568 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30987621 Lieferant BLD Pharm Lieferanten-Nr. BD1058771g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

(Stabilized with HQ)

Specifications

Chemischer Name oder Material N-Methylmethacrylamide
CAS 3887-02-3
Summenformel C5H9NO
MDL-Nummer MFCD00048115
Menge 1 g
SMILES C=C(C)C(NC)=O
Molekulargewicht (g/mol) 99.13
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.