missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-sulfonic Acid (Technical Grade), TRC

Artikelnummer. 30134324
Change view
Click to view available options
Menge:
5 g
10 g
50 g
Unit Size:
10 Gramm
5 Gramm
50 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30134324 50 g 50 Gramm
30006123 5 g 5 Gramm
30012241 10 g 10 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134324 Supplier Toronto Research Chemicals Supplier No. TRCN37850050G

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-47-2
Summenformel C10H8O3S
Molekulargewicht (g/mol) 208.23
Synonym 1-Naphthylsulfonic Acid, 1-Naphthalenesulfonic Acid, 1-Naphthalenesulfonic acid, 1-Naphthylsulfonic acid, 1-Sulfonaphthalene, HT 907, α-Naphthalenesulfonic acid
IUPAC-Name naphthalene-1-sulfonic acid
SMILES OS(=O)(=O)c1cccc2ccccc12

Specifications

Chemischer Name oder Material Naphthalene-1-sulfonic Acid (Technical Grade)
Menge 50 g
InChI-Formel InChI=1S/C10H8O3S/c11-14(12,13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H,11,12,13)
Formelmasse 208.0194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.