missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nbs1 Antibody (7E4A2), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-26297SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Nbs1 Monoclonal specifically detects Nbs1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Spezifikation
| Nbs1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:2000, Simple Western 1:50, Flow Cytometry 1:200 - 1:400, ELISA, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1-50 - 1:100, Immunohistochemistry-Paraffin, Knockout Validated | |
| ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
| Mouse | |
| 85 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
| 4683 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 α |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| 7E4A2 | |
| Unconjugated | |
| O60934 | |
| NBN | |
| Nbs1 Antibody (7E4A2) was made to partial recombinant human NBS1 (between residues 400-650) expressed in E. coli [Uniprot O60934] | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Precipitation |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur