missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NEK8, GST Tag Recombinant Protein
This NEK8 Recombinant Human Protein (1-322) was expressed in insect cells.
Marke: Invitrogen™ A30982
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
- This NEK8 recombinant human protein (1-322) was expressed in insect cells
- NEK8 serine/threonine kinase, or NIMA-related kinase 8, is related to NIMA (never in mitosis, gene A) of Aspergillus nidulans
- It may play a role in cell cycle progression from G2 to M phase
- NEK8 is part of a complex of ciliary proteins required for renal and cardiovascular development
- NEK8 interacts with polycystin signal transduction pathways and may control the targeting of these ciliary proteins
- NEK8 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Spezifikation
NP_835464 | |
62.3 kDa | |
NEKs | |
Human | |
GST Tag | |
Dry Ice | |
Recombinant, Active, Catalytic |
Kinase Assay | |
10 μg | |
aa 1 - 322 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |