Learn More
Invitrogen™ NGLY1 Recombinant Rabbit Monoclonal Antibody (9C10R7)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558062
Beschreibung
Immunogen sequence: MAAAALGSSS GSASPAVAEL CQNTPETFLE ASKLLLTYAD NILRNPNDEK YRSIRIGNTA FSTRLLPVRG AVECLFEMGF EEGETHLIFP KKASVEQLQK IRDLIAIERS SRLDGSNKSH KVKSSQQPAA STQLPTTPSS NPSGLNQHTR NRQGQSSDPP SASTVAADSA ILEVLQSNIQ HVLVYENPAL QEKALACIPV QELKRKSQEK LSRARKLDKG INISDEDFLL LELLHWFKEE FFHWVNNVLC SKCGGQTRSR DRSLLPSDDE LKWGAKEVED HYCDACQFSN RFPRYNNPEK.
This gene encodes an enzyme that catalyzes hydrolysis of an N(4)-(acetyl-beta-D-glucosaminyl) asparagine residue to N-acetyl-beta-D-glucosaminylamine and a peptide containing an aspartate residue. The encoded enzyme may play a role in the proteasome-mediated degradation of misfolded glycoproteins. Multiple transcript variants encoding different isoforms have been found for this gene.
Spezifikation
| NGLY1 | |
| Recombinant Monoclonal | |
| 0.67 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q96IV0, Q9JI78 | |
| NGLY1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human NGLY1 (NP_060767.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 9C10R7 | |
| Unconjugated | |
| NGLY1 | |
| 1110002C09Rik; CDDG; CDG1V; hPNGase; mPNGase; NGLY1; N-glycanase 1; peptide N-glycanase homolog; Peptide:N-glycanase; peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase; PNG1; PNGase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55768, 59007 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.