missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 7/CHRNA7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00 € - 326.00 €
Spezifikation
| Antigen | Nicotinic Acetylcholine R alpha 7/CHRNA7 |
|---|---|
| Verdünnung | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|
18605135
|
Novus Biologicals
NBP2-99069-100ul |
100 μg |
326.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18666905
|
Novus Biologicals
NBP2-99069-50ul |
50 μg |
227.00 €
50 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Description
Nicotinic Acetylcholine R alpha 7/CHRNA7 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 7/CHRNA7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Nicotinic Acetylcholine R alpha 7/CHRNA7 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 μm filtered solution in PBS | |
| 1139 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| a7 nicotinic acetylcholine receptor, alpha 7 neuronal nicotinic acetylcholine receptor, alpha-7 nicotinic cholinergic receptor subunit, cholinergic receptor, nicotinic, alpha 7, cholinergic receptor, nicotinic, alpha polypeptide 7, CHRNA7-2, NACHRA7, neuronal acetylcholine receptor protein, alpha-7 chain, neuronal acetylcholine receptor subunit alpha-7 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nicotinic Acetylcholine R alpha 7/CHRNA7. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title