missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitromethane, ≥99.0%, ReagentPlus™, Honeywell™
Click to view available options
Menge:
25 mL
100 mL
Spezifikation
Chemischer Name oder Material | Nitromethane |
Dichte | 1.127g/mL at 25°C |
Flammpunkt | 36°C (96.8°F) |
Menge | 25 mL |
UN-Nummer | UN1261 |
Beilstein | 1698205 |
Formelmasse | 61.04g/mol |
Reinheit (%) | ≥99.0% |
Güte | ReagentPlus™ |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'