Learn More
NMUR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14619-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Receptor for the neuromedin-U and neuromedin-S neuropeptides.Spezifikation
NMUR1 | |
Polyclonal | |
Unconjugated | |
NMUR1 | |
FM 3, FM3, G protein coupled receptor 66, GPC R, GPR66, neuromedin U receptor 1, NMUR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10316, 14767 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O55040, Q9HB89 | |
NMUR1 | |
NMUR1 Fusion Protein Ag6235 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |