Learn More
Invitrogen™ Novex™ APO-1/FAS/CD95 Human ELISA Kit
Quantifies human APO-1/FAS/CD95 protein in serum and tissue culture supernatant
Marke: Invitrogen™ KHS9501
Weitere Details : Netto-Gewicht : 0.70000kg
Beinhaltet: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
Beschreibung
APO-1 mediates apoptosis.
Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Immunoassays, Mitochondria, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Room Temperature
Spezifikation
<20pg/mL | |
Colorimetric Detection | |
355 | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays |
Plasma, Serum, Supernatant | |
Microplate Reader | |
2 hr. | |
APO-1 | |
Fas (TNF receptor superfamily, member 6) |
For Research Use Only. Not for use in diagnostic procedures.