Learn More
Invitrogen™ Luminex™ GM-CSF Human Singleplex Bead Kit
Analyte specific components for the measurement of human GM-CSF (Granulocyte-Macrophage Colony Stimulating Factor) in serum, plasma or tissue culture supernatant
Marke: Invitrogen™ LHC2011
Weitere Details : Netto-Gewicht : 0.05000kg
Beinhaltet: Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet.
Benötigt : Buffer reagents needed to complete the reaction sold separately (LHB0001).
Beschreibung
The assay may be run alone or in combination with other Human Singleplex Bead Kits. EveryPrep™ Universal Vacuum Manifold available separately (Cat. No. K2111-01). Reagents intended for use in Luminex 100™ or 200™ Systems only.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Wet Ice
Spezifikation
100 Assays | |
<15 pg/mL | |
Luminex Instruments | |
NM_000758.3, NP_000749.2 | |
Cell Culture Supernatant, Plasma, Serum |
Polystyrene | |
Fluorescent Detection | |
1437 | |
colony stimulating factor 2 (granulocyte-macrophage) | |
Capture antibody coated beads, Detector antibody, Lot specific technical data sheet, Standard |
For Research Use Only. Not for use in diagnostic procedures.