Learn More
Invitrogen™ Novex RBP4 Competitive Human ELISA Kit
Designed to quantify the levels of RBP (Retinol binding protein) 4 in serum, plasma, and cell culture supernatants
Marke: Invitrogen™ KHP0081
Weitere Details : Netto-Gewicht : 0.50000kg
Beinhaltet: Pre-coated plate, standard, control, detector antibody, anti-rabbit HRP, diluents, wash buffer, chromogen, and stop solution.
Beschreibung
RBP4 is an adipocyte-derived hormone that is a transport protein for vitamin A with a novel role in regulating insulin action.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Bestellinformation
Shipping Condition: Wet ice
Spezifikation
Plasma, Serum, Supernatant | |
5950 | |
1 ng/mL | |
For research use only (RUO) | |
2.3 hr. |
Colorimetric | |
anti-rabbit HRP, Chromogen, Control, Detector antibody, Diluents, Pre-coated plate, Standard, Stop solution, Wash buffer | |
96 Assays | |
Retinol binding protein 4, plasma |
For Research Use Only. Not for use in diagnostic procedures.