Learn More
NOX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 17772-1-AP-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16877744
  • 145.00 € / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Three splice variants of NOX1 have been identified, NOH-1L, NOH-1S and NOH-1Lv. NOH-1S is a voltage-gated proton channel that participates in the regulation of cellular pH and is blocked by zinc. NOH-1L is a pyridine nucleotide-dependent oxidoreductase that generates superoxide and might conduct H (+) ions as part of its electron transport mechanism, whereas NOH-1S does not contain an electron transport chain. NOX1 have the potential to be effective treatments for a range of ischemic diseases.
Spezifikation
Spezifikation
NOX1
Polyclonal
Unconjugated
NOX1
GP91 2, Mitogenic oxidase 1, MOX 1, MOX1, NADPH oxidase 1, NOH 1, NOH1, NOX 1, NOX1
Rabbit
Antigen Affinity Chromatography
RUO
114243, 237038, 27035
-20°C
Liquid
Western Blot, ELISA
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8CIZ9, Q9WV87, Q9Y5S8
Nox1
NOX1 Fusion Protein Ag12187
20 μL
Primary
Mouse, Rat, Human