missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noxa Antibody (114C307.1R) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-07012
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Noxa Monoclonal antibody specifically detects Noxa in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular)
Spezifikation
| Noxa | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 μg/ml, Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1-5 μg/ml, Flow (Intracellular) | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
| 0.1 mg | |
| Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Mitochondrial Mediated Pathway, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| 5366 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 114C307.1R | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur