missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noxa Antibody (114C307.1R) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
267.75 € - 549.15 €
Spezifikation
| Antigen | Noxa |
|---|---|
| Klon | 114C307.1R |
| Konzentration | 1.0 mg/mL |
| Verdünnung | Western Blot 1 - 2 μg/ml, Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1-5 μg/ml, Flow (Intracellular) |
| Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18641617
|
Novus Biologicals
NBP3-07012 |
0.1 mg |
581.00 € 549.15 € / 0.10 Milligramm Sparen 31.85 € 5% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18631887
|
Novus Biologicals
NBP3-07012SS |
0.025 mg |
284.00 € 267.75 € / 25 Mikrogramm Sparen 16.25 € 5% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Noxa Monoclonal antibody specifically detects Noxa in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Flow (Intracellular)Spezifikation
| Noxa | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Mitochondrial Mediated Pathway, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| PBS | |
| 5366 | |
| IgG1 κ | |
| Protein A or G purified |
| 114C307.1R | |
| Western Blot 1 - 2 μg/ml, Flow Cytometry, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1-5 μg/ml, Flow (Intracellular) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts