missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSDHL Recombinant Rabbit Monoclonal Antibody (24GB485)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557523
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NSDHL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.
Spezifikation
| NSDHL | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Nsdhl | |
| AI747449; bare patches; Bpa; H105E3; NAD(P) dependent steroid dehydrogenase-like; NAD(P)-dependent steroid dehydrogenase-like protein; Nsdhl; Protein H105e3; SDR31E1; short chain dehydrogenase/reductase family 31E, member 1; sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; Str; striated; XAP104 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 50814 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB485 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15738 | |
| Nsdhl | |
| A synthesized peptide derived from human NSDHL (aa 175-200). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur