missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NSDHL Recombinant Rabbit Monoclonal Antibody (24GB485)

Rabbit Recombinant Monoclonal Antibody
204.00 € - 580.00 €
Spezifikation
| Antigen | NSDHL |
|---|---|
| Klon | 24GB485 |
| Inhalt und Lagerung | -20°C |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30276227
![]() |
Invitrogen™
MA557522 |
100 μL |
580.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30276404
|
Invitrogen™
MA557523 |
20 μL |
204.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
NSDHL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.Spezifikation
| NSDHL | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Nsdhl | |
| AI747449; bare patches; Bpa; H105E3; NAD(P) dependent steroid dehydrogenase-like; NAD(P)-dependent steroid dehydrogenase-like protein; Nsdhl; Protein H105e3; SDR31E1; short chain dehydrogenase/reductase family 31E, member 1; sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; Str; striated; XAP104 | |
| Nsdhl | |
| Primary | |
| Affinity chromatography |
| 24GB485 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15738 | |
| 50814 | |
| A synthesized peptide derived from human NSDHL (aa 175-200). | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts