Learn More
Duncan & Associates™ Nylongaze
Artikelnummer. 10467752
Encompass
Ansicht ändern
Click to view available options
Breite (metrisch):
1.2 mm
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Breite (metrisch) Packungsgröße
10467752 1.2 mm 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 10467752 Lieferant Duncan & Associates™ Lieferanten-Nr. GMX500V
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
missing translation for 'validMainframeMsg'
Spezifikation
Produkttyp Nylongaze
Abmessungen (L x B) 0.5 x 1.2 mm
Menge 1/Packung
Länge (metrisch) 0.5 mm
Material Nylon
Breite (metrisch) 1.2 mm