missing translation for 'onlineSavingsMsg'
Learn More

O-Allylhydroxylamine Hydrochloride, 97%, BLD Pharm™

Artikelnummer. 30986857
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30986857 1 g
30985505 5 g
30981850 500 g
30984390 100 g
30970895 25 g
30982059 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30986857 Lieferant BLD Pharm Lieferanten-Nr. BD1459281g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material O-Allylhydroxylamine Hydrochloride
CAS 38945-21-0
Summenformel C3H8ClNO
MDL-Nummer MFCD00012957
Menge 1 g
SMILES NOCC=C.[H]Cl
Molekulargewicht (g/mol) 109.55
Reinheit (%) 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.