missing translation for 'onlineSavingsMsg'
Learn More

O-Desmorpholinopropyl-O-methyl-gefitinib, TRC

Product Code. 17931335
Change view
Click to view available options
Menge:
10 mg
50 mg
100 mg
Unit Size:
10 Milligramm
100 Milligramm
50 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
17931335 50 mg 50 Milligramm
30136563 100 mg 100 Milligramm
30135483 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17931335 Supplier Toronto Research Chemicals Supplier No. TRCD29684050MG
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153437-78-6
Summenformel C16H13ClFN3O2
Molekulargewicht (g/mol) 333.74
Synonym 4-Quinazolinamine, N-(3-chloro-4-fluorophenyl)-6,7-dimethoxy-, N-(3-Chloro-4-fluorophenyl)-6,7-dimethoxy-4-quinazolinamine, N-(3-Chloro-4-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine
IUPAC-Name N-(3-Chlor-4-Fluorophenyl)-6,7-Dimethoxyquinazolin-4-Amin
SMILES COc1cc2ncnc(Nc3ccc(F)c(Cl)c3)c2cc1OC

Specifications

Chemischer Name oder Material O-Desmorpholinopropyl-O-methyl-gefitinib
Menge 50 mg
InChI-Formel InChI=1S/C16H13ClFN3O2/c1-22-14-6-10-13(7-15(14)23-2)19-8-20-16(10)21-9-3-4-12(18)11(17)5-9/h3-8H,1-2H3,(H,19,20,21)
Formelmasse 333.068
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.