Learn More
OCSTAMP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556648
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RLQRRHDRHQ GQQLPLGDPS CVPTPRPACK PPAWIDYRLD ALRTESSEGE GKELWSCRDL SCNLGPVPPP CVTLGKSLHL SEPRFLHLHN DSIFTIDVTY FPRRDVVRME GNTGHD Highest antigen sequence identity to the following orthologs: Mouse - 20%, Rat - 20%.
OCSTAMP Polyclonal antibody specifically detects OCSTAMP in Human samples. It is validated for Immunohistochemistry.Spezifikation
OCSTAMP | |
Polyclonal | |
Unconjugated | |
OCSTAMP | |
C20orf123; dJ257E24.3; OCSTAMP; OC-STAMP; Osteoclast stimulatory transmembrane protein; transmembrane protein C20orf123 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128506 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BR26 | |
OCSTAMP | |
Recombinant protein corresponding to Human OCSTAMP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only