missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 (OCT3) Mouse, anti-Human, PE, Clone: 3A2A20, STEMCELL Technologies
Mouse Monoclonal Antibody
318.60 € - 711.45 €
Spezifikation
| Antigen | OCT4 (OCT3) |
|---|---|
| Klon | 3A2A20 |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Konjugat | PE |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17188971
|
Stemcell Technologies
60093PE.1 |
25 Tests |
318.60 €
25 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
17178971
|
Stemcell Technologies
60093PE |
100 Tests |
711.45 €
100 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
- The 3A2A20 antibody reacts with human OCT4 (octamer-binding transcription factor 4; also known as OCT3 and OCT3/4), an ∼40 kDa homeodomain transcription factor belonging to the POU family, which is expressed in undifferentiated human embryonic stem (ES), induced pluripotent stem (iPS), embryonal carcinoma (EC) and embryonic germ (EG) cells
- OCT4 binds to the octamer motif (5'-ATTTGCAT-3') and plays a key role in maintaining cells in a pluripotent state by interacting with other transcription factors such as SOX2 to regulate the expression of several genes, including FBX15, FGF-4, REX1, SOX2 and osteopontin
- Levels of OCT4 are down-regulated during differentiation and it has thus emerged as a useful marker of pluripotency in stem cells, as well as a marker for certain human malignant germ cell tumours
- Expression of OCT4 together with other transcription factors has been used to reprogram somatic cells into iPS cells
Spezifikation
| OCT4 (OCT3) | |
| Flow Cytometry, Western Blot, Immunocytochemistry, Immunofluorescence | |
| PE | |
| Human | |
| Primary |
| 3A2A20 | |
| Monoclonal | |
| Mouse | |
| IgG2b κ |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.
Name des Produkts