missing translation for 'onlineSavingsMsg'
Learn More

Kern Sohn OIML Class M1 Weight Set, Plastic Case

Artikelnummer. 17392213
Encompass_Preferred
Click to view available options
Gewicht (metrisch):
1 bis 100 g
1 bis 200 g
1 bis 50 g
1 bis 500 g
1 g bis 1 kg
1 g bis 2 kg
1 g bis 5 kg
1 mg bis 100 g
1 mg bis 1 kg
2 mg bis 1 kg
5 mg bis 1 kg
Packungsgröße:
1 Set
1 Stück
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17392213

Marke: Kern Sohn 344024

Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Weight set constructed of finely turned stainless steel and housed in a plastic case.

  • Knob shape
  • Finely turned stainless steel
  • Lined case
  • Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic box

Specifications

Klassifikation M1
Material Stainless Steel
Gehäusematerial Plastic
Gewicht (metrisch) 1 bis 50 g
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.