missing translation for 'onlineSavingsMsg'
Learn More

Oligodendrocyte Marker O4 Mouse, anti-Human, Unconjugated, Clone: 81, STEMCELL Technologies

Artikelnummer. 17138771
Encompass_Preferred
Change view
Click to view available options
Menge:
50 μg
Product Code. Menge
17138771 50 μg
1 options
This item is not returnable. View return policy

Product Code. 17138771

Brand: Stemcell Technologies 60053

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

  • O-Antigens are expressed on oligodendrocytes during differentiation and on mature oligodendrocytes in the central nervous system
  • O4 is formed during postnatal development and is a marker for cell bodies and processes of oligodendrocytes types I and II
  • O4 is expressed in pro-oligodendrocytes, however, it is not found on O-2A-progenitor cells

Specifications

Antigen Oligodendrocyte Marker O4
Anwendungen Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Klassifikation Monoclonal
Klon 81
Konjugat Unconjugated
Wirtsspezies Mouse
Menge 50 μg
Primär oder sekundär Primary
Zielspezies Human, Mouse, Pig, Rat
Isotype IgM
Zelltyp Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.