Learn More
OMG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 720194
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Bovine and Cat.
OMG Polyclonal antibody specifically detects OMG in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
OMG | |
Polyclonal | |
Unconjugated | |
OMG | |
oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; OMGP | |
Rabbit | |
Affinity chromatography | |
RUO | |
4974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
P23515 | |
OMG | |
Protein corresponding to Human OMG (aa 26-130). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |