Learn More
ONECUT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563393
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MLTRGEQHLS RGLGTPPAAM MSHLNGLHHP GHTQSHGPVL APSRERPPSS SSGSQVATSG QLEE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
ONECUT2 is a member of the transcription factors of the ONECUT class, whose prototype is hepatocyte nuclear factor (HNF)-6. The distribution of OC-2 mRNA in humans is tissue-restricted, the strongest expression being detected in the liver and skin. ONECUT2 participates in the network of transcription factors required for liver differentiation and metabolism.Spezifikation
ONECUT2 | |
Polyclonal | |
Unconjugated | |
ONECUT2 | |
HNF-6B, OC-2, one cut domain family member 2, OC2, OC-2, one cut homeobox 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95948 | |
ONECUT2 | |
Recombit protein corresponding to Human ONECUT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only