missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR1E1/OR1E2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00253W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Glycerol (1:1) may be added for additional stability.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Spezifikation
OR1E1/OR1E2 | |
Polyclonal | |
OR1E2 | |
Olfactory receptor 1E1, Olfactory receptor 13-66, OR13-66, Olfactory receptor 17-2/17-32, OR17-2, OR17-32, Olfactory receptor 1E5, Olfactory receptor 1E6, Olfactory receptor 5-85, OR5-85, Olfactory receptor OR17-18, Olfactory receptor-like protein HGMP07I, OR1E1, OR1E5, OR1E6, OR1E9P, Olfactory receptor 1E2, Olfactory receptor 17-93/17-135/17-136, OR17-135, OR17-136, OR17-93, Olfactory receptor 1E4, OR1E2, OR1E4 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P30953, P47887 | |
OR1E1, OR1E2 | |
A synthetic peptide from human olfactory receptor | |
RUO | |
8387, 8388 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |