missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2B11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561525
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SDNHSFLGDS PKAFILLGVS D Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 62%.
Odorant receptor.Spezifikation
OR2B11 | |
Polyclonal | |
Unconjugated | |
OR2B11 | |
novel 7 transmembrane receptor (rhodopsin family) protein; olfactory receptor 2B11; olfactory receptor family 2 subfamily B member 11; olfactory receptor, family 2, subfamily B, member 11; OR2B11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127623 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JQS5 | |
OR2B11 | |
Recombinant protein corresponding to Human OR2B11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only